Rat Recombinant IL17A
Supplier: IBI Scientific
Certificates
About this item
IBI recombinant proteins are expressed in a yeast system (Pichia pastoris) which promotes protein folding and post-translational modifications for greater bio-activity. IBI proteins are Endotoxin FREE and are purified without HIS-TAGS or an other TAG. Our yeast expression system has proven to produce more active proteins that more closely resemble their host form.
IBI proteins are lyophilized and can be shipped at room temperature. Proteins can be reconstituted in sterile PBS with at least 0.1% of carrier protein such as BSA or cell assay media.
The recombinant rat protein IL-17A has a gene ID of 301289. The calculated molecular weight is 15.0kDa.
Specifications
- Protein/peptide type:Recombinant
- Species:Rat
- Gene ID:301289
- Endotoxin-free:Yes
- Protein/peptide name:IL17A
Specifications
Frequently Bought Together