Order Entry
Canada
ContactUsLinkComponent
Rat Recombinant APRIL
Rat Recombinant APRIL
Supplier:  IBI Scientific
Certificates
Rat Recombinant APRIL
Supplier:  IBI Scientific

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Rat
  • Gene ID:
    287437
  • Endotoxin-free:
    Yes
  • Protein/peptide name:
    APRIL

Specifications

Frequently Bought Together