KIMBLE® BEVEL-SEAL™ Connecting Adapters, DWK Life Sciences
KT179920-0808
- Tubing ext.Ø:8 to 8 mm O.D.
- Description:BEVEL-SEAL® PTFE Connecting Adapter with FKM 109 O-ring and Open Top Polypropylene Cap
- Environmentally Preferable:
- Cat. no.:KT179920-0808
- O-Ring Size:109
- Supplier No.:179920-0808
Connects to vacuum systems, NMR tubes, photochemical cuvettes, lyophilizing tubes, flowmeters, and more.
- Simple o-ring connections
- Vacuum seal
- Body is made from PTFE
Simple o-ring connections form a vacuum seal on glass tubing by tightening the compression cap. The body is PTFE.
: Supplied with FKM o-ring and open top polypropylene cap.