Metallocene Liners, Associated Bag
Clear, 22×16×59", 3 mil.
47751-122
: 247-6-53C
This new-generation polyethylene resin offers more strength and tear resistance than low density poly which is 25% thicker.
Use with 50 to 55 gallon cans; circumference measures 76".
- Color:Clear
- Thickness:3 mil.
- W×L:22×16×59"
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



