Williams Medium E, Quality Biological
Catalog # 10128-636
Supplier: QUALITY BIOLOGICAL, INC.
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Williams medium E without L-Glutamine
- Size:500 ml
- Sterility:Sterile filtered
- Shipping conditions:Ambient temperature
- pH:7.0 ±0.2
- Cat. no.:10128-636
- Shelf Life:24 months
- Supplier No.:112-033-101
Specifications
About this item
Dr. Williams and his coworkers’ studies on hepatocytes in culture led to the development of Williams’ Medium E.
- Without L-glutamine
This rich medium was used to culture the hepatocytes derived from an enrichment procedure also developed by Dr. Williams and his colleagues in 1971. The work on hepatocytes from the newborn led to attempts to culture adult liver on a long term basis. Williams’ Medium E has unique components that make it a reduced serum-supplemented medium.
Sterile Filtered
Harmonization Code: 3821.00.0000
ECCN: EAR99
It is recommended to add L-glutamine to a final concentration of 2.0mM.
Williams and his coworkers' studies on hepatocytes in culture led to the development of Williams' Medium E.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research Use Only