SharpsMaster II® 9014 Gloves, HexArmor
Supplier: HexArmor
Certificates
About this item
Sharpmaster II 9014 provide the highest needlestick resistance available, by layering SuperFabric brand material.
- Tested using actual 25 gauge needles against ANSI/ISEA 105 standard
- Highest level of cut resistance on noted enhanced areas, ANSI/ISEA Cut Level A9 and Needlestick Level 5
- Single glove needle solution with incredible dexterity and comfort
- Wrinkle rubber palm coating with Actifresh™ anti-microbial treatment
HexArmor sets the industry standard for needle protection with innovative solutions for a variety of applications. The Sharpsmaster II® 9014 is a manufacturing breakthrough that utilizes high performance SuperFabric® brand material technology engineered to provide the highest levels of needle and cut protection available on the market. This glove gives end users incredible protection in a single glove solution. Don't get stuck with faulty hand protection - become a Sharpsmaster.
Specifications
- Catalog No:
- 76222-506
- 76222-504
- 10147-432
- 10147-430
- 10147-428
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Elastic
- Elastic
- Elastic
- Elastic
- Elastic