ContactUsLinkComponent
E.Z.N.A.® Micro RNA Kits, Omega Bio-Tek
E.Z.N.A.® Micro RNA Kits, Omega Bio-Tek
Catalog # CA11006-516
Supplier:  Omega Bio-Tek
CAS Number:  
E.Z.N.A.® Micro RNA Kits, Omega Bio-Tek
Catalog # CA11006-516
Supplier:  Omega Bio-Tek
Supplier Number:  R7034-01
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    E.Z.N.A.® Micro RNA Kit
  • Size:
    50 Preps
  • Cat. no.:
    CA11006-516
  • Supplier No.:
    R7034-01
Specifications
About this item
The E.Z.N.A
The system uses RNA-Solv to efficiently lyse cells and protect RNA from degradation. Following a chloroform separation step, RNA is applied to a HiBind RNA column where large RNA binds. The flow through is retained and applied to a HiBind Micro RNA column to bind small RNA.
Micro RNA Kit combines the reversible binding properties of HiBind® matrix, a new silica-based material, with the highly efficient lysis ability of RNA-Solv® to extract micro and large RNA from a wide variety of starting materials. Micro and large RNA are separated to allow for quantification into downstream applications.