Calf Native Alkaline Phosphatase (from Intestine)
Supplier: MilliporeSigma
Certificates
About this item
Native Alkaline Phosphatase from calf intestine is a homodimeric enzyme that hydrolyzes phosphate esters or primary and secondary alcohols, amines and phenols
It is useful in hydrolysis of 5'-terminal phosphates of DNA and RNA.
Specifications
- Enzyme type:Native
- Source:Intestine
- Species:Calf
- Enzyme name:Alkaline Phosphatase
- Enzyme synonyms:AP
Specifications
Frequently Bought Together