Order Entry
Canada
ContactUsLinkComponent
Anti-SELL Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEL-14]
Anti-SELL Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEL-14]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-SELL Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MEL-14]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10050-940
  • 10050-936
  • 10050-938
  • Antigen symbol:
  • SELL
  • SELL
  • SELL
  • Antigen name:
  • Selectin L
  • Selectin L
  • Selectin L
  • Antigen synonyms:
  • LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
  • LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
  • LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • MEL-14
  • MEL-14
  • MEL-14
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Gene ID:
  • 20343
  • 20343
  • 20343
  • Isotype:
  • IgG2a
  • IgG2a
  • IgG2a
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Yes
  • Storage temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together