Order Entry
Canada
ContactUsLinkComponent
 
Anti-PTPRC Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 104]
 
undefined
Anti-PTPRC Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 104]

 

  • Catalog No:
  • 10050-922
  • 10050-918
  • 10050-920
  • Antigen symbol:
  • PTPRC
  • PTPRC
  • PTPRC
  • Antigen name:
  • Protein Tyrosine Phosphatase Receptor Type C
  • Protein Tyrosine Phosphatase Receptor Type C
  • Protein Tyrosine Phosphatase Receptor Type C
  • Antigen synonyms:
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • 104
  • 104
  • 104
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Gene ID:
  • 19264
  • 19264
  • 19264
  • Isotype:
  • IgG2a
  • IgG2a
  • IgG2a
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Yes
  • Storage temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry
  • Flow Cytometry

 

Frequently Bought Together