About this item
The DTA-1 antibody reacts with mouse CD357, also known as GITR or AITR (in humans), a 66-70 kDa member of the Tumor Necrosis Factor superfamily (TNFRSF18). GITR is primarily found on T cells, and its function may vary depending on the T cell type where it is expressed. GITR is upregulated on activated T cells where it provides co-stimulation, yet GITR may promote the inhibition of CD4+ CD25+ Treg cells, where it is expressed at high levels. GITR ligand (GITRL) is found on B cells, macrophages, dendritic and endothelial cells, and is implicated in regulating both innate and adaptive immune responses.
The DTA-1 antibody may be used for analysis of GITR expression on T cells, and is also commonly used in vitro as an agonistic antibody to induce GITR signaling in various assays.
Type: Primary
Antigen: TNFRSF18
Clonality: Monoclonal
Clone: DTA-1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Specifications
- Catalog No:
- 10051-006
- 10051-008
- Antigen symbol:
- TNFRSF18
- TNFRSF18
- Antigen name:
- Tumor Necrosis Factor Receptor Superfamily Member 18
- Tumor Necrosis Factor Receptor Superfamily Member 18
- Antigen synonyms:
- CD357|TNFRSF18|Activation-inducible TNFR Family Receptor|Glucocorticoid-induced TNFR-related Protein|AITR|Tumor Necrosis Factor Receptor Superfamily Member 18
- CD357|TNFRSF18|Activation-inducible TNFR Family Receptor|Glucocorticoid-induced TNFR-related Protein|AITR|Tumor Necrosis Factor Receptor Superfamily Member 18
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- DTA-1
- DTA-1
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 21936
- 21936
- Isotype:
- IgG2b
- IgG2b
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Storage temperature:
- Store undiluted at 2-8°C, protect from light, do not freeze.
- Store undiluted at 2-8°C, protect from light, do not freeze.
- Tested applications:
- Flow Cytometry
- Flow Cytometry