Order Entry
Canada
ContactUsLinkComponent
Mouse IgG2b Isotype Control [clone: MPC-11] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Mouse IgG2b Isotype Control [clone: MPC-11] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Catalog # 10141-208
Supplier:  Tonbo Biosciences
undefined
Mouse IgG2b Isotype Control [clone: MPC-11] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Catalog # 10141-208
Supplier:  Tonbo Biosciences
Supplier Number:  65-4732-U100

Specifications

  • Antibody type:
    Control
  • Antigen name:
    Mitochondrial pyruvate carrier 11
  • Clone:
    MPC-11
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Size:
    100 µg
  • Storage temperature:
    Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested applications:
    Flow Cytometry
  • Cat. no.:
    10141-208
  • Supplier No.:
    65-4732-U100

Specifications

About this item

The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.

Type: Control
Antigen: MPC11
Clonality:
Clone: MPC-11
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: