Mouse IgG2b Isotype Control [clone: MPC-11] (PE (Phycoerythrin)-Cy7®)
10141-060
- Antibody type:Control
- Antigen name:Mitochondrial pyruvate carrier 11
- Clone:MPC-11
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Size:100 µg
- Storage temperature:Store undiluted at 2-8°C, protect from light, do not freeze.
- Tested applications:Flow Cytometry
- Cat. no.:10141-060
- Supplier No.:60-4732-U100
The MPC-11 immunoglobulin is useful as an isotype-matched control. The MPC-11 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2b antibodies.
Type: Control
Antigen: MPC11
Clonality:
Clone: MPC-11
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: