Order Entry
Canada
ContactUsLinkComponent
Anti-CD2 Rat Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RM2-5]
Anti-CD2 Rat Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RM2-5]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD2 Rat Antibody (FITC (Fluorescein Isothiocyanate)) [clone: RM2-5]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10141-146
  • 10141-144
  • 10141-148
  • Antigen symbol:
  • CD2
  • CD2
  • CD2
  • Antigen name:
  • CD2 molecule
  • CD2 molecule
  • CD2 molecule
  • Antigen synonyms:
  • sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
  • sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
  • sheep red blood cell receptor|T-cell surface antigen T11|erythrocyte receptor|rosette receptor|T11|Leu-5|SRBC|LFA-3 receptor|lymphocyte-function antigen-2|LFA-2
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • RM2-5
  • RM2-5
  • RM2-5
  • Reactivity:
  • Mouse
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Rat
  • Gene ID:
  • 12481
  • 12481
  • 12481
  • Isotype:
  • IgG2b
  • IgG2b
  • IgG2b
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Storage temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together