Order Entry
Canada
ContactUsLinkComponent
Anti-IL4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11B11]
Anti-IL4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11B11]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-IL4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11B11]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10050-660
  • 10050-658
  • Antigen symbol:
  • IL4
  • IL4
  • Antigen name:
  • Interleukin 4
  • Interleukin 4
  • Antigen synonyms:
  • BCGF-1|B-cell stimulatory factor 1|IL-4|Interleukin 4|BCGF1|BSF1|BSF-1|Interleukin-4
  • BCGF-1|B-cell stimulatory factor 1|IL-4|Interleukin 4|BCGF1|BSF1|BSF-1|Interleukin-4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 11B11
  • 11B11
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16189
  • 16189
  • Isotype:
  • IgG1
  • IgG1
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Shipping temperature:
  • Store undiluted at 2...8 °C, protect from light, do not freeze.
  • Store undiluted at 2...8 °C, protect from light, do not freeze.
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together