Order Entry
Canada
ContactUsLinkComponent
Anti-LPAM-1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DATK32]
Anti-LPAM-1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DATK32]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-LPAM-1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: DATK32]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10051-350
  • 10051-352
  • Antigen symbol:
  • LPAM-1
  • LPAM-1
  • Antigen synonyms:
  • CD49d/beta7|alpha4beta7 Integrin|LPAM-1
  • CD49d/beta7|alpha4beta7 Integrin|LPAM-1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • DATK32
  • DATK32
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Isotype:
  • IgG2a
  • IgG2a
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Shipping temperature:
  • Store undiluted at 2...8 °C, protect from light, do not freeze.
  • Store undiluted at 2...8 °C, protect from light, do not freeze.
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together