ContactUsLinkComponent
Nunc™ Screwdriver for Cryobank and Bank-It™ Vial Systems, Thermo Scientific
Nunc™ Screwdriver for Cryobank and Bank-It™ Vial Systems, Thermo Scientific
Catalog # CA82030-534
Nunc™ Screwdriver for Cryobank and Bank-It™ Vial Systems, Thermo Scientific
Catalog # CA82030-534
Supplier Number:  369643
Specifications
  • Description:
    Manual Screwdriver, black
  • Cat. no.:
    CA82030-534
  • Supplier No.:
    369643
Specifications
About this item
Quickly and easily cap and decap Nunc Cryobank tubes using the manual Nunc™ Screwdriver for Cryobank and Bank-It™ Systems
Nunc Cryobank vials provide the customer with superior tracking of their cryovials using a 2D code, the highest storage density on the market and the ability to automate the capping and decapping of the vials. The tubes click into the rack and can be handled without the risk of accidentally dropping a vial.