Orders LinkContactUsLinkComponent
Adapt-A-Rack® Multi-Tube Racks
Adapt-A-Rack® Multi-Tube Racks
Supplier:   Heathrow Scientific
Adapt-A-Rack® Multi-Tube Racks
Certificates
Adapt-A-Rack® Multi-Tube Racks
Adapt-A-Rack® Multi-Tube Racks
8 Options Available Below
Product Details & Documents
Adapt-A-Rack® are capable of holding different sized tubes in each available slot as the well literally adapts to hold the contents firmly in an upright position.
  • Made of POM
  • Racks can be linked together for additional holding capacity
  • Open sides allow for label viewing
  • Flat base to hold tubes upright
  • Available in solid or a combination of colors for easy coding identification
  • Autoclavable
They can be linked together for additional holding capacity. Multiple colors can be combined to create a larger rack system. Each rack will accommodate four tubes.
Product Family Options
Product Information
  • Color
    No. of Holes
    For tube O.D.
    For tubes
    Dimensions
    Availability
    Price
  • Blue
    4
    12 - 30 mm
    7 - 50 ml
    181×56×76 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $37.29
    $37.29
    New Customer Promotional Price
    Log in for availability
    Case of 20
    Case of 20 - $372.88
    $372.88
    New Customer Promotional Price
    Pack of 2
       $37.29
    Case of 20
       $372.88
    Supplier Number: HS120187
  • Blue/Green Combo
    4+4
    12 - 30 mm
    7 - 50 ml
    181×56×76 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $37.81
    $37.81
    New Customer Promotional Price
    Log in for availability
    Case of 20
    Case of 20 - $378.08
    $378.08
    New Customer Promotional Price
    Pack of 2
       $37.81
    Case of 20
       $378.08
    Supplier Number: HS120184
  • Blue/Yellow Combo
    4+4
    12 - 30 mm
    7 - 50 ml
    181×56×76 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $37.81
    $37.81
    New Customer Promotional Price
    Log in for availability
    Case of 20
    Case of 20 - $378.08
    $378.08
    New Customer Promotional Price
    Pack of 2
       $37.81
    Case of 20
       $378.08
    Supplier Number: HS120185
  • Green
    4
    12 - 30 mm
    7 - 50 ml
    181×56×76 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $37.29
    $37.29
    New Customer Promotional Price
    Log in for availability
    Case of 20
    Case of 20 - $372.88
    $372.88
    New Customer Promotional Price
    Pack of 2
       $37.29
    Case of 20
       $372.88
    Supplier Number: HS120191
  • Pink
    4
    12 - 30 mm
    7 - 50 ml
    181×56×76 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $37.57
    $37.57
    New Customer Promotional Price
    Log in for availability
    Case of 20
    Case of 20 - $375.73
    $375.73
    New Customer Promotional Price
    Pack of 2
       $37.57
    Case of 20
       $375.73
    Supplier Number: HS120189
  • Pink/White Combo
    4+4
    12 - 30 mm
    7 - 50 ml
    181×56×76 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $37.29
    $37.29
    New Customer Promotional Price
    Log in for availability
    Case of 20
    Case of 20 - $372.88
    $372.88
    New Customer Promotional Price
    Pack of 2
       $37.29
    Case of 20
       $372.88
    Supplier Number: HS120186
  • White
    4
    12 - 30 mm
    7 - 50 ml
    181×56×76 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $37.81
    $37.81
    New Customer Promotional Price
    Log in for availability
    Case of 20
    Case of 20 - $378.08
    $378.08
    New Customer Promotional Price
    Pack of 2
       $37.81
    Case of 20
       $378.08
    Supplier Number: HS120188
  • Yellow
    4
    12 - 30 mm
    7 - 50 ml
    181×56×76 mm
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 2
    Pack of 2 - $37.67
    $37.67
    New Customer Promotional Price
    Log in for availability
    Case of 20
    Case of 20 - $376.66
    $376.66
    New Customer Promotional Price
    Pack of 2
       $37.67
    Case of 20
       $376.66
    Supplier Number: HS120190