Order Entry
Canada
ContactUsLinkComponent
Anti-IL2RA Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: BC96]
Anti-IL2RA Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: BC96]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-IL2RA Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: BC96]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10051-476
  • 10051-478
  • Antigen symbol:
  • IL2RA
  • IL2RA
  • Antigen name:
  • Interleukin 2 Receptor, Alpha
  • Interleukin 2 Receptor, Alpha
  • Antigen synonyms:
  • TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
  • TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)-Cy7®
  • PE (Phycoerythrin)-Cy7®
  • Clone:
  • BC96
  • BC96
  • Reactivity:
  • Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Baboon, Cynomolgus monkey, Macaque, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3559
  • 3559
  • Isotype:
  • IgG1
  • IgG1
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage temperature:
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Store undiluted at 2-8°C, protect from light, do not freeze.
  • Shipping temperature:
  • Store undiluted at 2...8 °C, protect from light, do not freeze.
  • Store undiluted at 2...8 °C, protect from light, do not freeze.
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together