About this item
The ICRF44 antibody reacts with human CD11b, also known as integrin alpha M. This 165-170 kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alpha M (CD11b) assembles with Integrin beta-2 (CD18) into a receptor known as Macrophage Antigen-1 (Mac-1) or complement receptor type 3 (CR3). This receptor binds and induces intracellular signaling through ICAM-1, ICAM-2, ICAM-3 and ICAM-4 on endothelial cells and can also facilitate removal of iC3b bearing foreign cells.
The ICRF44 antibody is widely used as a marker for CD11b expression on macrophages, granulocytes, and subsets of NK cells. It is reported to be cross-reactive with a number of non-human species including Baboon, Chimpanzee, Cynomolgus, Rhesus and Swine.
Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: ICRF44
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Baboon, Chimpanzee, Cynomolgus, Rhesus, Pig
Specifications
- Catalog No:
- 10050-880
- 10050-882
- Antigen symbol:
- ITGAM
- ITGAM
- Antigen name:
- Integrin Alpha M
- Integrin Alpha M
- Antigen synonyms:
- CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
- CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- ICRF44
- ICRF44
- Reactivity:
- Baboon, Pig, Cynomolgus monkey, Macaque, Chimpanzee, Human
- Baboon, Pig, Cynomolgus monkey, Macaque, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3684
- 3684
- Isotype:
- IgG1
- IgG1
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Storage temperature:
- Store undiluted at 2-8°C, protect from light, do not freeze.
- Store undiluted at 2-8°C, protect from light, do not freeze.
- Tested applications:
- Flow Cytometry
- Flow Cytometry