About this item
HyClone™ donor horse serum, designed to supplement cell growth, is collected from selected donor herds that receive regular veterinary inspection and care.
- Horses are bled in an enclosed facility that is specially equipped for aseptic collection
- Coggins tests for equine infectious anemia are routinely performed on each horse
- Serum filtered using sequential 100 nm pore-size rated filters
Supports hybridoma and antibody production. Meets specifications of \<=10EU/mL of endotoxin and \<=10mg/dL of hemoglobin. Filtered through triple 100nm pore-size rated filtration system. A biochemical profile is supplied.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA16777-026
- CA16777-030
- CA82013-590
- Storage temperature:
- Less than –10 °C
- Less than –10 °C
- Less than –10 °C
Specifications
Related Information
Certificates
Search by Lot NumberCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
HyClone™ Cell Culture Serum products