Order Entry
Canada
ContactUsLinkComponent
Hydrochloric acid 6.00 N
Hydrochloric acid 6.00 N
Catalog # RC3750.1-5PT
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Hydrochloric acid 6.00 N
Catalog # RC3750.1-5PT
Supplier:  Ricca Chemical
Supplier Number:  3750.1-5PT
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, clear
  • Size:
    2.5 L
  • Cat. no.:
    RC3750.1-5PT
  • Supplier No.:
    3750.1-5PT

Specifications

Specification Test Results

  • Assay:
    6 ± 0.005 N at 20 °C
  • NIST SRM:
    84

Specification Test Results

About this item

 :
ADR:8,II
 :36.46
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324