Order Entry
Canada
ContactUsLinkComponent
Hydrochloric acid 3.00 N NF, USP
Hydrochloric acid 3.00 N NF, USP
Catalog # R3720100-4A
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
Hydrochloric acid 3.00 N NF, USP
Catalog # R3720100-4A
Supplier:  Ricca Chemical
Supplier Number:  R3720100-4A
Small Business Enterprise
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Poly natural
  • Size:
    4 L
  • Cat. no.:
    R3720100-4A
  • Supplier No.:
    R3720100-4A

Specifications

Specification Test Results

  • Assay:
    3 ± 0.003 N at 20 °C
  • NIST SRM:
    84

Specification Test Results

About this item

 :
ADR:8,II
 :36.46
UN:1789
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324