Order Entry
Canada
ContactUsLinkComponent
EDTA disodium salt 0.0500 M (M/20) in aqueous solution ASTM D 2036 D titrant
EDTA disodium salt 0.0500 M (M/20) in aqueous solution ASTM D 2036 D titrant
Catalog # RC2705-16
Supplier:  Ricca Chemical
Small Business Enterprise
CAS Number:  
undefined
EDTA disodium salt 0.0500 M (M/20) in aqueous solution ASTM D 2036 D titrant
Catalog # RC2705-16
Supplier:  Ricca Chemical
Supplier Number:  2705-16
Small Business Enterprise
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Size:
    500 mL
  • Cat. no.:
    RC2705-16
  • Supplier No.:
    2705-16

Specifications

Specification Test Results

  • Assay:
    0.05 ± 0.00001 M at 20 °C
  • NIST SRM:
    915

Specification Test Results

About this item

 :
 :336.21 g/mol
Boiling Pt:100 °C
Melting Pt:0.0 °C
Density:1.0090 g/cm³
Storage Temperature:Ambient
CAS Number:139-33-3
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672