About this item
HyClone USDA-tested Fetal Bovine Serum (FBS) provides important proteins, hormones, growth factors, metabolites, and nutrients for your cell culture processes at a competitive price. USDA-tested FBS is ideal for typical cell culture research applications.
- Complete traceability back to original source
- Low in antibodies and high in growth factors
- Produced using true pool technology for uniformity and consistency from bottle to bottle
- Filtered through three sequential 100 nm (0,1 µm) pore size-rated filters
- Sourced from countries recognized as being low risk of BSE and FMD by the USDA
- Once thawed, sera should be stored at 2 to 8 °C
USDA-Tested Fetal bovine serum (FBS) is sourced from bovine spongiform encephalopathy (BSE)-negligible regions in throughout the world for use in low-risk applications. This serum is filtered in the US and considered to be foot and mouth disease (FMD)-free.
Each lot is safety tested by the U.S. Department of Agriculture (USDA) in accordance with safety protocols required for the serum country of origin. This serum is largely sourced from Central America.
Additional treatments available including heat-inactivation and irradiation. Additional testing, including EMEA-conforming testing, is also available.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA89133-096
- CA89133-094
- CA76324-916
- CA89133-100
- CA89133-098
- Endotoxin:
- ≤25 EU/ml
- ≤25 EU/ml
- ≤25 EU/ml
- ≤25 EU/ml
- ≤ 25 EU/ml
- Hemoglobin:
- ≤25 mg/dl
- ≤25 mg/dl
- ≤25 mg/dl
- ≤25 mg/dl
- ≤25 mg/dl
- Storage buffer:
- Sterile filtered
- Storage temperature:
- Less than 10 °C
- Less than 10 °C
- Less than 10 °C
- Less than 10 °C
- Less than 10 °C
Specifications
Related Information
Certificates
Search by Lot NumberCytiva BioXtra and Pall Life Sciences Products CA Cytiva BioXtra and Pall Life Sciences Products US
Promotion Material
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
HyClone™ Cell Culture Serum products