Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Size:100 mL
- Storage temperature:Store Undiluted at 2 to 8°C
- Cat. no.:10141-140
- Supplier No.:TNB-4300-L100
- Reactivity:Mouse, Human
Specifications
About this item
RBC Lysis Buffer (10X) is a concentrated ammonium chloride-based lysing reagent
The diluted 1X working solution will lyse red blood cells in single cell suspensions with minimal effects on leukocytes. RBC Lysis Buffer (10X) does not contain a fixative so the cells remain viable after red blood cell lysis.