Anti-FOXP3 Rat Antibody (PE (Phycoerythrin)) [clone: MF23]
Supplier: Tonbo Biosciences
Certificates
About this item
The MF23 antibody reacts with mouse Foxp3, a 50-55 kDa transcription factor which is a central regulator of T cell activity and is critical for the development and function of regulatory T cells (Tregs). Foxp3 is a member of the forkhead box or winged helix family of transcription factors and is expressed at constitutively high levels in Treg cells. Forced expression of Foxp3 in conventional T cells results in the upregulation of Treg associated molecules such as CD25, CTLA-4, and GITR and is sufficient to impart suppressive functional activity to these cells.
Type: Primary
Antigen: Foxp3
Clonality:
Clone: MF23
Conjugation: PE
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Specifications
- Catalog No:
- 10141-198
- 10141-200
- Antigen symbol:
- FOXP3
- FOXP3
- Antigen name:
- Forkhead Box P3
- Forkhead Box P3
- Antigen synonyms:
- AIID|PIDX|JM2|DIETER|XPID|Scurfin|IPEX
- AIID|PIDX|JM2|DIETER|XPID|Scurfin|IPEX
- Antibody type:
- Primary
- Primary
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- MF23
- MF23
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 20371
- 20371
- Isotype:
- IgG2b
- IgG2b
- Flow cytometry:
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Storage temperature:
- Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested applications:
- Flow Cytometry
- Flow Cytometry
Specifications
Frequently Bought Together