Rat IgG1 Isotype Control [clone: HRPN]
Catalog # 10141-174
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Control
- Clone:HRPN
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Size:100 µg
- Neutralisation:Yes
- Storage temperature:Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested applications:Functional Assay
- Cat. no.:10141-174
- Supplier No.:40-4301-U100
Specifications
About this item
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.
Type: Control
Antigen: HRPN
Clonality:
Clone: HRPN
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: