Order Entry
Canada
ContactUsLinkComponent
Somso® Labyrinth with Ossicles and Tympanic Membrane
Somso® Labyrinth with Ossicles and Tympanic Membrane
Catalog # 470102-368
Supplier:  MARCUS SOMMER
undefined
Somso® Labyrinth with Ossicles and Tympanic Membrane
Catalog # 470102-368
Supplier:  MARCUS SOMMER
Supplier Number:  DS 17

Specifications

  • Description:
    Labyrinth with ossicles and tympanic membrane
  • Height:
    20 cm
  • Length:
    16 cm
  • Cat. no.:
    470102-368
  • Width:
    14 cm
  • Supplier No.:
    DS 17

Specifications

About this item

Somso® premium quality models.

  • Five year warranty

Enlarged approx. four times, in SOMSO-Plast®. The model clearly shows the organs of the middle ear space and the inner ear. The membranous and bony labyrinths are shown and the cochlea can be opened. Separates into three parts. On a stand with green base.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.