Order Entry
Canada
Orders LinkContactUsLinkComponent
Somso® Brain

Somso® Brain

8 part brain model, 15 cm, 18 cm, 15 cm
Catalog # 470102-306
Supplier:   MARCUS SOMMER
Supplier #:  BS 20
Somso® Brain
Catalog # 470102-306
Supplier:  MARCUS SOMMER
Somso® Brain
Somso® Brain

8 part brain model, 15 cm, 18 cm, 15 cm

Price
$1,290.00
Log in for availability
Each
Product Details & Documents

Somso® premium quality models.

  • Five year warranty

Natural cast, in SOMSO-Plast®. Separates into 8 parts: frontal and parietal lobes (2), temporal and occipital lobes (2), medulla (2), cerebellum (2). On a base.

Ordering information: Items on this page are not stocked and may have an extended time for delivery.

Specifications
  • Description:
    8 part brain model
  • Height:
    15 cm
  • Length:
    18 cm
  • Width:
    15 cm