Order Entry
Canada
ContactUsLinkComponent
 
Anti-ITGAM Rat Monoclonal Antibody (APC (Allophycocyanin)-Cy7®) [clone: M1/70]
 
undefined
Anti-ITGAM Rat Monoclonal Antibody (APC (Allophycocyanin)-Cy7®) [clone: M1/70]

 

  • Catalog No:
  • 10050-694
  • 10050-692
  • Antigen symbol:
  • ITGAM
  • ITGAM
  • Antigen name:
  • Integrin Alpha M
  • Integrin Alpha M
  • Antigen synonyms:
  • CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
  • CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)-Cy7®
  • APC (Allophycocyanin)-Cy7®
  • Clone:
  • M1/70
  • M1/70
  • Reactivity:
  • Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Mouse
  • Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16409 / 3684
  • 16409 / 3684
  • Isotype:
  • IgG2b
  • IgG2b
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage temperature:
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

 

Frequently Bought Together