Mouse IgG2a Isotype Control [clone: C1.18.4] (PE (Phycoerythrin)-Cy7®)
10141-058
- Antibody type:Control
- Clone:C1.18.4
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Size:100 µg
- Storage temperature:Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested applications:Flow Cytometry
- Cat. no.:10141-058
- Supplier No.:60-4724-U100
The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.
Type: Control
Antigen: C1.18.4
Clonality:
Clone: C1.18.4
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: 0