Mouse IgG2a Isotype Control [clone: C1.18.4]
Catalog # 10141-180
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Control
- Clone:C1.18.4
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Size:100 µg
- Neutralisation:Yes
- Storage temperature:Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested applications:Functional Assay
- Cat. no.:10141-180
- Supplier No.:40-4724-U100
Specifications
About this item
The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.
Type: Control
Antigen: C1.18.4
Clonality:
Clone: C1.18.4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: 0