Order Entry
Canada
ContactUsLinkComponent
Mouse IgG2a Monoclonal Isotype Control [clone: C1.18.4] (FITC (Fluorescein Isothiocyanate))
Mouse IgG2a Monoclonal Isotype Control [clone: C1.18.4] (FITC (Fluorescein Isothiocyanate))
  10050-986
undefined
Mouse IgG2a Monoclonal Isotype Control [clone: C1.18.4] (FITC (Fluorescein Isothiocyanate))
  10050-986
 :  35-4724-U100

 

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    C1.18.4
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Size:
    100 µg
  • Neutralisation:
    Yes
  • Storage temperature:
    Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Tested applications:
    Flow Cytometry
  • Cat. no.:
    10050-986
  • Supplier No.:
    35-4724-U100

 

 

The C1.18.4 immunoglobulin is useful as an isotype-matched control. The C.1.18.4 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG2a antibodies.

Type: Control
Antigen: C1.18.4
Clonality: Monoclonal
Clone: C1.18.4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: 0