Mouse IgG1 Isotype Control [clone: MOPC-21] (redFluor® 710)
100 µg
Catalog # 10141-256
Supplier: Tonbo Biosciences
Supplier #: 80-4714-U100
Mouse IgG1 Isotype Control [clone: MOPC-21] (redFluor® 710)
Catalog # 10141-256
Supplier: Tonbo Biosciences
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.
Type: Control
Antigen: MOPC21
Clonality:
Clone: MOPC-21
Conjugation: redFluor® 710
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Documents
Specifications
- Size:100 µg
- Cat. No.:10141-256
- Flow cytometry:Yes
- Antibody type:Control
- Neutralisation:Yes
- Isotype:IgG1
- Storage temperature:Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Host:Mouse
- Conjugation:redFluor® 710
- Tested applications:Flow Cytometry
- Clone:MOPC-21
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



