Order Entry
Canada
ContactUsLinkComponent
Anti-IFNG Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: XMG1.2]
Anti-IFNG Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: XMG1.2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-IFNG Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: XMG1.2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10051-042
  • 10051-044
  • Antigen symbol:
  • IFNG
  • IFNG
  • Antigen name:
  • Interferon, Gamma
  • Interferon, Gamma
  • Antigen synonyms:
  • AF1|IFG|Interferon gamma transducerInterferon gamma|IFI|Interferon gamma receptor |IFN-gamma|Immune interferon
  • AF1|IFG|Interferon gamma transducerInterferon gamma|IFI|Interferon gamma receptor |IFN-gamma|Immune interferon
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • XMG1.2
  • XMG1.2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 15978
  • 15978
  • Isotype:
  • IgG1
  • IgG1
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage temperature:
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together