Specifications
- Antibody type:Primary
- Antigen name:Killer Cell Lectin-Like Receptor Subfamily B, Member 1
- Antigen symbol:KLRB1C
- Clonality:Monoclonal
- Clone:PK136
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Size:100 µg
- Neutralisation:Yes
- Gene ID:17059
- Antigen synonyms:NK-RP1|ly-55c|Nk1|AI462337|NKR-P1.9|Ly-59|Nk-1.2|NK1.1|Ly55c|Ly59|Nk-1|Nkrp1c|NKRP140|CD161|Nk1.2|NKRP1
- Storage temperature:Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested applications:Immunofluorescence Microscopy
- Cat. no.:10051-732
- Supplier No.:70-5941-U100
Specifications
About this item
The PK136 antibody is specific for mouse NK1.1, a type II transmembrane lectin-like receptor and member of the killer cell lectin-like receptor (KLR) family. NK1.1 is prominently expressed on natural killer (NK) cells, and is correlated with NK cytotoxic effects toward virus-infected cells and tumor cells. NK1.1 is expressed on subsets of NKT cells in certain mouse strains (C57BL/6, FVB/N, and NZB), yet absent from others (AKR, BALB/c, CBA/J, C3H, DBA/1, DBA/2, NOD, SJL, and 129). Putative subsets of NK cells and their expression of NK1.1 antigen are of continuing interest, including NK1.1+/CD117+ (c-Kit) cells reported to be immunosuppressive for CD8+ T cells in a mechanism involving PD-1 and PD-L1 (Ehlers et al. 2012. Endocrinology. 10: 1247).
The PK136 antibody may be used for detection of NK1.1 expression on mouse strains including CE, B6, NZB, C58, Ma/My, ST, SJL, and FVB. The antibody is reported to react with an epitope common to NKR-P1B and NKR-P1C alloantigenic forms of NK1.1 (Carlyle et al. 2006. J. Immunol. 176: 7511-7524).
Type: Primary
Antigen: KLRB1C
Clonality: Monoclonal
Clone: PK136
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse