Order Entry
Canada
ContactUsLinkComponent
Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: RM4-5]
Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: RM4-5]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD4 Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: RM4-5]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 10051-434
  • 10051-432
  • Antigen symbol:
  • CD4
  • CD4
  • Antigen name:
  • CD4 molecule
  • CD4 molecule
  • Antigen synonyms:
  • L3T4|p55|CD4mut|Ly-4|W3/25
  • L3T4|p55|CD4mut|Ly-4|W3/25
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)-Cy7®
  • PE (Phycoerythrin)-Cy7®
  • Clone:
  • RM4-5
  • RM4-5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12504
  • 12504
  • Isotype:
  • IgG2a
  • IgG2a
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Storage temperature:
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together