Orders LinkContactUsLinkComponent
Vernier® Electrode Support
Vernier® Electrode Support
Vernier electrode support
Catalog # CA470112-920
New Product
Supplier #:  ESUP
Vernier® Electrode Support
Catalog # CA470112-920
New Product
Vernier® Electrode Support
Vernier® Electrode Support
Vernier electrode support
Price
$18.09
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Our Electrode Support is a great complement to the Vernier Stir Station, as well as a perfect holder for many sensors. It is built to connect to all standard ring-stand posts; its large-handled locking nut keeps your sensors firmly in place.
  • The larger opening (13 mm Ø) is perfect for our pH, ISE, Conductivity and ORP sensors
  • The smaller opening (5 mm) is designed to hold our Stainless Steel Temperature Probe or the Vernier Go!Temp
Specifications
  • Description:
    Vernier electrode support