Orders LinkContactUsLinkComponent
Vernier® Diffraction Apparatus
Vernier® Diffraction Apparatus
Vernier diffraction apparatus, Light sensor full scale ranges: 1, 10, and 100 µW; Linear position sensor range: 150 mm
Catalog # CA470112-972
New Product
Supplier #:  DAK
Vernier® Diffraction Apparatus
Catalog # CA470112-972
New Product
Vernier® Diffraction Apparatus
Vernier® Diffraction Apparatus
Vernier diffraction apparatus, Light sensor full scale ranges: 1, 10, and 100 µW; Linear position sensor range: 150 mm
Price
$1,435.09
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Map light intensity vs. position for various slit geometries.
  • Red laser wavelength 635 nm ±5 nm (Class 2)
  • 40 µm linear position sensor wavelength
The diffraction apparatus lets students create, view, and measure diffraction and interference patterns. The included red diffraction laser provides a clean monochromatic light source. Slits are made by depositing metal film on glass, creating extraordinarily clean slits with fully opaque blocking areas. These high-precision slits cast clean diffraction and interference patterns, ideal for quantitative matching of intensity vs. position predictions.
The linear position sensor and high sensitivity light sensor completes the diffraction apparatus. In order to provide excellent spatial resolution, a selectable entrance aperture restricts the width of the pattern viewed by the High Sensitivity Light Sensor. The light sensor has three ranges, allowing the study of fine details or gross features of patterns. To measure position, the light sensor is mounted on a novel Linear Position Sensor.
This position sensor uses a precision optical encoder to measure translation with better than 50-micron resolution. Since it is optically based, without gears or racks, it has zero backlash.
Data collection is performed by choosing a slit, directing the laser through the slit, and choosing an entrance aperture for the light sensor. Both the light sensor and position sensor connect to your interface. The light sensor is moved by hand over a 150 mm distance, tracing out the diffraction pattern.
Ordering information: LabQuest® (2, 3, Mini) interface is required but not included. 1.2 m Track/optics bench available individually or as part of the dynamics cart and track system. An optional green diffraction laser is also available separately.
Delivery information: Includes diffraction slit system, red diffraction laser, combination linear position and high sensitivity light sensor, and power supply.
Specifications
  • Description:
    Vernier diffraction apparatus
  • Range:
    Light sensor full scale ranges: 1, 10, and 100 µW; Linear position sensor range: 150 mm