You searched for: Gaskets
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Supplier: VWR International
Smart Gasket lets you take samples or readings from a sanitary line.
Expand 35 Items
VWR® Vacuum Oven Gasket
Supplier: VWR International
For use with VWR® Vacuum Ovens.
Expand 1 Items
Masterflex® Smart Gasket® Sanitary Gasket Accessories, Avantor®
Supplier: VWR International
Optimize performance of your Masterflex® Smart Gasket® Sanitary Gasket system.
Expand 2 Items
Masterflex® Tuf-Steel® Sanitary Tri-Clamp® Gaskets, Avantor®
Supplier: VWR International
The benefits of PTFE and 316L stainless steel in one gasket!
Expand 6 Items
Masterflex® Sanitary Gaskets, Polymers, Avantor®
Supplier: VWR International
Ensure a tight seal.
Expand 33 Items
Masterflex® Flanged Sanitary Gaskets, Tri-Clamp®, Avantor®
Supplier: VWR International
Hands-free installation for difficult-to-reach locations!
Expand 3 Items
Masterflex® Sanitary Tri-Clamp® Screen Gaskets, Viton, Avantor®
Supplier: VWR International
Protect your pump, valves and other components in fluid line.
Expand 21 Items
Masterflex® Tuf-Flex® Sanitary Tri-Clamp® Gaskets, Avantor®
Supplier: VWR International
Ensure a better process seal with outstanding chemical resistance.
Expand 7 Items
Masterflex® Sanitary Tri-Clamp® Sock Screen Gaskets, Avantor®
Supplier: VWR International
Greater particulate capacity for higher particulate flow.
Expand 24 Items
Masterflex® Sanitary Tri-Clamp® Screen Gaskets, PTFE, Avantor®
Supplier: VWR International
Protect your pump, valves and other components in fluid line.
Expand 20 Items
Masterflex® Single-Use Sanitary Gaskets, TPE, Avantor®
Supplier: VWR International
Ensure a tight seal in your sanitary fluid line.
Expand 3 Items
Masterflex® X-Rayable and Metal Detectable Sanitary Gaskets, Avantor®
Supplier: VWR International
Easily detect lost elastomeric fragments to prevent contamination and product recalls.
Expand 9 Items
Masterflex® Smart Gasket® 3-Port Sanitary Gaskets, Silicone, Avantor®
Supplier: VWR International
Easily sample fluid in critical high-purity system.
Expand 1 Items
Masterflex® Smart Gasket® 1-Port Sanitary Gaskets, Silicone, Avantor®
Supplier: VWR International
Easily sample fluid in critical high-purity system.
Expand 1 Items
Gauge Guard Protector and Isolator Seals
Supplier: VWR International
Extend the life and accuracy of your expensive and fragile instruments.
Expand 4 Items
Masterflex® Platinum-cured Silicone Screen Gaskets, Avantor®
Supplier: VWR International
Protect your pump, valves and other components in fluid line.
Expand 28 Items
Masterflex® Pressure Gauge Guard Gaskets and Protector Caps, Avantor®
Supplier: VWR International
Simple accessories to protect your delicate pressure gauge.
Expand 10 Items
Masterflex® Smart Gasket® Sanitary Clamps, Stainless Steel, Avantor®
Supplier: VWR International
Create a secure connection with your Masterflex® Smart Gasket® sanitary gaskets.
Expand 16 Items
Masterflex® Sanitary Tri-Clamp® Screen Gaskets, EPDM, Avantor®
Supplier: VWR International
Protect your pump, valves and other components in fluid line.
Expand 23 Items
Masterflex® Tuf-Steel® Sanitary Screen Tri-Clamp Gaskets, Avantor®
Supplier: VWR International
Protect your pump, valves and other components in fluid line.
Expand 13 Items
Gaskets, NW Flange, Ace Glass Incorporated
Supplier: Ace Glass
NW (DN) flange glass to glass sealing gasket featuring a PTFE carrier and o-ring available in Viton® (–26 to 240 °C) or FVMQ flurosilicone (–60 to 230 °C).
Expand 6 Items
Nalgene® Sanitary Gaskets for Nalgene Carboys; Silicon, Thermo Scientific
Supplier: Thermo Fisher Scientific
Sanitary gaskets for Nalgene carboys are recommended for use with 2630 and 2640 carboys series.
Expand 3 Items
Intuvo Gaskets, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Features and specifications for Intuvo Gaskets.
Expand 3 Items
Metal to Glass Gasket, NW Flange, Ace Glass
Supplier: Ace Glass
NW (DN) flange metal to glass sealing gasket featuring a PTFE carrier and O-ring.
Expand 6 Items
Glass to Glass Gasket, NW Flange, Ace Glass
Supplier: Ace Glass
NW (DN) flange glass to glass sealing gasket featuring a PTFE carrier and O-ring.
Expand 3 Items
Gasket for Ground Flanges, Ace Glass Incorporated
Supplier: Ace Glass
Gaskets are designed for mating ground flange surfaces on reactor flasks and matching heads.
Expand 5 Items
Metal to Metal Connection, NW Flange, Ace Glass
Supplier: Ace Glass
Connections for NW metal flanges to like NW metal flanges.
Expand 8 Items
Round Gasket with Rectangular Hole, Bioptechs Inc.®
Supplier: BIOPTECHS INC.
A round silicone gasket featuring a rectangular 30 mm I.D.
Expand 1 Items
Gasket, Perfluoro, Black, 400 mm DBR Industries
Supplier: Chemglass
Class VI Perfluoroelastomer with 3 tabs each spaced 120 degrees apart along the outer edge to aid in gasket alignment.