Orders LinkContactUsLinkComponent
Casters for Modular Workstations
Casters for Modular Workstations
Supplier:   Treston
Casters for Modular Workstations
Supplier:  Treston
Certificates
Casters for Modular Workstations
Casters for Modular Workstations
1 Options Available Below
Product Details & Documents
Make any Sovella lab bench a mobile lab station by adding casters! Work is not always confined to one location.
  • Contain foot activated total lock brake
  • Easy swiveling and smooth rolling
  • Durable zinc plated finish
Ordering information: Casters sold in sets of four. Installation is required. Assembly videos available.
Product Family Options
Product Information
  • Load capacity
    Availability
    Price
  • 89202-112
    Low profile casters lower worksurfaces
    Low profile casters lower worksurfaces
    200 lbs.
    Discontinued
    Supplier Number: -
  • 89202-110
    Lab caster set of 4 (2 locking, 2 non-locking)
    Lab caster set of 4 (2 locking, 2 non-locking)
    660 lbs.
    Log in for availability
    Log in for availability
    Each
    Each - $365.97
    $365.97
    Each
       $365.97
    Minimum order quantity is 1
    Supplier Number: 834734-00
  • 89202-114
    Roll and set casters, set of 4
    Roll and set casters, set of 4
    1100 lbs.
    Discontinued
    Supplier Number: -
  • Discontinued
    Supplier Number: -
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.