Orders LinkContactUsLinkComponent
Sash-Glider Kit, Labconco
Sash-Glider Kit, Labconco
Sash-Glider Kit
Catalog # 10127-516
Supplier:   Labconco
Supplier #:  3408500
Sash-Glider Kit, Labconco
Catalog # 10127-516
Supplier:  Labconco
Sash-Glider Kit, Labconco
Sash-Glider Kit, Labconco
Sash-Glider Kit
Price
$336.00
Log in for availability
Kit of 1
Minimum order quantity is 1
Product Details & Documents
Use the Sash-Glider to clean both sides of a biological safety cabinet's sash while it remains closed, to avoid contamination risks.
Delivery information: Sash-Glider Kit includes magnetic outside and inside blades, 2 microfiber sheets and 10 disposable white scrubbing strips. Additional scrubbing strips are available.
Specifications
  • Description:
    Sash-Glider Kit
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Refill Pack of 25 Sash-Glider Scrubbing Strips
    Refill Pack of 25 Sash-Glider Scrubbing Strips
    10127-518
    Log in for availability
    Kit of 25 - $289.00