STAR™ DISH Petri Dishes with Markings
Supplier: Phoenix Biomedical
Certificates
About this item
These Petri dishes are ideal for antibiotic susceptibility testing
They accommodate two to three membrane filters per dish, and they are large enough for bioburden determination and sterility testing of medical devices. The extremely flat bottom has markings for placing up to six strips of Etest®.
Frequently Bought Together