Conventional Petri Dishes, Square with Grid, Phoenix Biomedical
Catalog #: CA25389-326
Supplier: Phoenix Biomedical
Specifications
- Dimensions:100 x 15 mm
- Description:Square Petri Dish with Grid
- Cat. no.:CA25389-326
- Supplier No.:1050-050
Specifications
About this item
These square petri dishes feature 13 mm grid marks in both directions
Dishes are suitable for bacteriological typing, probe assays, colony counts, susceptibility testing or any research or bacterial culture work requiring a large surface area.
Customers Who Bought This Also Bought