Conventional Petri Dishes, Phoenix Biomedical
Catalog # CA25389-048
Supplier: Phoenix Biomedical
Specifications
- Description:Petri Dish without Vents, with ISO Mark
- Dimensions:90 x 15 mm
- Cat. no.:CA25389-048
- Supplier No.:113
Specifications
About this item
This dish offers distortion-free viewing, making it ideal for microscopic examination
The controlled bottom flatness ensures even distribution of media and organisms. The semi-stacking ring has been specifically designed to make the dish versatile and suitable for different makes and models of pouring equipment.