R-Phycoerythrin (RPE), Columbia Biosciences
Catalog # 10065-816
Supplier: Columbia Biosciences
CAS Number:
Specifications
- Description:R-Phycoerythrin (RPE)
- Size:1 mg
- Storage temperature:Store at 4° C
- Application:Flow Cytometry, Cell-Based Assays, Microarrays and Microplate Applications
- Ex/Em:565-498/578 nm
- Cat. no.:10065-816
- Dye:SureLight® R-PE
- Supplier No.:D5-000
Specifications
About this item
Many applications and instruments were developed specifically for R-phycoerythrin
It is commonly used in immunoassays such as FACS, flow cytometry, multimer/tetramer applications. With new instrumetation available, R-PE is also well suited for immunohistochemistry (IHC) and Western blot applications.