ContactUsLinkComponent
SP Bel-Art Stopcocks, Bel-Art Products, a part of SP
SP Bel-Art Stopcocks, Bel-Art Products, a part of SP
Catalog # 59097-102
undefined
SP Bel-Art Stopcocks, Bel-Art Products, a part of SP
Catalog # 59097-102
Supplier Number:  F42040-0000
Specifications
  • Description:
    Replacement Stopcocks
  • Cat. no.:
    59097-102
  • Fits Tubing I.D.:
    ¼ - ⅜"
  • Supplier No.:
    420400000
Specifications
About this item
Polypropylene plastic body has Teflon® PTFE plug, washer and bright red handle.
Taper fits on desiccator port, and the plug is 3 way for vacuum draw, shut-off and vacuum release.