Orders LinkContactUsLinkComponent
Specimen Container, Dynalon
Specimen Container, Dynalon
Supplier:   Dynalon
Specimen Container, Dynalon
Supplier:  Dynalon
Certificates
Specimen Container, Dynalon
Specimen Container, Dynalon
6 Options Available Below
Product Details & Documents
Lightweight containers with tight-fitting lids are non-breakable and translucent
Designed to simplify and standardize the handling and storage of specimens. The containers are inert and cannot be attacked by formaldehyde, weak acids, or bases. They will resist freezing or boiling. Specimen containers offer long shelf lives and are easily disposed, yet sturdy enough for reuse. Easy to store, containers require little space. Dimension values include container with lid.
Product Family Options
Product Information
  • Capacity
    Height
    Weight
    Material
    Ø
    Availability
    Price
  • 240 ml (7.7 oz.)
    3.5"
    0.028 lbs.
    PPCO
    3.1"
    Log in for availability
    Log in for availability
    Case of 250
    Case of 250 - $502.69
    $502.69
    Case of 250
       $502.69
    Supplier Number: 454405
  • 480 ml (15.5 oz.)
    4.0 "
    0.052 lbs.
    PPCO
    3.6"
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $252.81
    $252.81
    Case of 100
       $252.81
    Supplier Number: 454415
  • 960 ml (34.5 oz.)
    5.3 "
    0.09 lbs.
    PPCO
    4.6"
    Log in for availability
    Log in for availability
    Case of 100
    Case of 100 - $388.53
    $388.53
    Case of 100
       $388.53
    Supplier Number: 454425
  • 2000 ml (67.6 oz.)
    6.6 "
    0.152 lbs.
    HDPE
    5.9"
    Log in for availability
    Log in for availability
    Case of 50
    Case of 50 - $374.53
    $374.53
    Case of 50
       $374.53
    Supplier Number: 454435
  • 2200 ml (83.6 oz.)
    6.6"
    0.18 lbs.
    PPCO
    6.4"
    Log in for availability
    Log in for availability
    Case of 25
    Case of 25 - $333.18
    $333.18
    Case of 25
       $333.18
    Supplier Number: 453555
  • 4800 ml (168.8 oz.)
    7.8"
    0.374 lbs.
    HDPE
    8.1"
    Log in for availability
    Log in for availability
    Case of 25
    Case of 25 - $375.02
    $375.02
    Case of 25
       $375.02
    Supplier Number: 454445
  • 0.028 lbs.
    Discontinued
    Supplier Number: -