Order Entry
Canada
ContactUsLinkComponent
 
Human Recombinant SCF (from E. coli)
 :  Prosci
 
Human Recombinant SCF (from E. coli)
 :  Prosci

 

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Biological activity:
    The ED50 was determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is = 2.0 ng/mL, corresponding to a specific activity of = 5 x 105 units/mg.
  • Protein synonyms:
    Kit ligand|SCF|KL-1|Kitl|SF|FPH2|Mast cell growth factor|MGF|SHEP7
  • Protein/peptide name:
    SCF
  • Purity:
    >98%
  • Formulation:
    Lyophilized
  • Gene ID:
    60427

 

Frequently Bought Together